Full Scale Human X-Rays | Avantor
Full Scale Human X-Rays
Full Scale Human X-Rays
Catalog # 470007-886
Supplier:   Roylco
Fournisseur #:  R5911
Full Scale Human X-Rays
Catalog # 470007-886
Supplier:  Roylco
Full Scale Human X-Rays
Full Scale Human X-Rays
Full Scale Human X-Rays
Price
$71.05
Log in pour la disponibilité
KIT(1items) Kit of 1 Case of 24
Product Details & Documents
The entire skeleton of a 5' tall person.
  • Computer-enhanced images
  • Individual panels
  • Information sheets included
Computer-enhanced and printed onto heavy-duty plastic sheets, this set of 18 real human x-rays is an excellent visual aid for anatomy studies. The individual panels can be arranged together to reproduce the entire skeleton of a 5' tall young adult. Study x-rays on a light table, overhead projector, or held up to a window. Includes information sheet in English and French with reproducible diagrams for labeling. X-ray size range: 7 3/8" x 1 1/4" to 13 3/4" x 11".
Spécifications
  • Désignation:
    Full Scale Human X-Rays