Ward's® Frog Development Poster
Frog Development Poster, Unlaminated, 86 cm, 53 cm
Catalog # 470023-248
Supplier: Avantor
Jump to:
Product Details & Documents
Developmental Stages Portrayed with Eye-Catching Photos
- Students will gain a deeper appreciation of the complexity of the amphibian life cycle
Your students will gain a deeper appreciation of the complexity of the amphibian life cycle by studying this poster. Color photos detail the many stages of frog development, from egg to tadpole to frog.
Spécifications
- Largeur:53 cm
- Désignation:Frog Development Poster
- Longueur:86 cm
- Finish:Unlaminated