Freeze-Dried Grassfrogs
Supplier: Avantor
Certificates
About this item
Freeze-dried grassfrogs. Available in vacuum packs.
- Amphibian anatomy
- Economical
Our freeze-dried specimens, which use no chemicals, can be rehydrated in 10% alcohol for dissection.
Frequently Bought Together