Four-Channel Timer-Stopwatch with Clock
Four-channel timer
Catalog # 470001-920
Supplier: General Tools Manufacturing
Fournisseur #: TI388
Jump to:
Product Details & Documents
This timer features four independent countdown timers-stopwatches that count up to or down from 99:59:59.
12/24-hour clock
Counts up after counting down to zero
Remembers the last timer setting
A magnet, flip-up stand and belt clip are on the back
It will automatically count up after the countdown reaches zero. Alarm duration is 1 minute. LCD size is 1.88×0.88" (48×22mm), with 5/8" (16mm) high digits. Power source is a "LR44" battery (included). A 1 year limited warranty is also included.
.
Spécifications
- Désignation:Four-channel timer