Forward Vision II Workstation
Forward Vision II Workstation
   Sustainable Options Available
Forward Vision II Workstation
   Sustainable Options Available
Certificates
Forward Vision II Workstation
2 Options Available Below
Product Details & Documents
The science teacher´s workstation is designed to be used by four students, all facing the instructor. It is compact enough to fit multiple units in a small classroom and is easy to assemble.
  • Large ¾" thick chemical resistant phenolic resin work surface
  • Constructed of solid oak and oak veneers
  • Rubber base molding and durable chemical resistant UV finish
  • Full cabinet doors with locks include adjustable shelves
  • Side mounted hooks hold book bags off the floor.
  • Two GFI protected AC duplex receptacles and data outlets on the side of the unit
Available with flat top or solid epoxy sink with two combination cold water and gas fixtures.
Max. load: 500 lbs. per unit; 80 lbs. per shelf
Certifications: SEFA compliant and MAS Certified Green.
Specifications for Product Family
Specifications
  • Catalog No:
  • 470117-678
  • 470117-676
  • Environmentally Preferable:
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Environmental certification:
  • MAS Certified Green
  • MAS Certified Green
Specifications
Product Family Options
Product Information
  • Désignation
    Capacité
    No. of Users
    Worksurface Color/Finish
    Worksurface Material
    Worksurface Thickness
    Hauteur
    W×D
    Weight
    Availability
    Price
  • Forward vision II workstation, with fixtures
    500 lbs.
    5
    Black
    Phenolic resin
    ¾"
    36"
    88×46"
    503 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $12,799.90
    $12,799.90
    Each
       $12,799.90
    Minimum order quantity is 1
    Supplier Number: 2944K
    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    470117-678
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    MAS Certified Green
      Safer Human and Environmental Health
  • Forward vision II workstation, flat top
    500 lbs.
    5
    Black
    Phenolic resin
    ¾"
    36"
    88×46"
    503 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $10,285.80
    $10,285.80
    Each
       $10,285.80
    Minimum order quantity is 1
    Supplier Number: 2944KF
    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    470117-676
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    MAS Certified Green
      Safer Human and Environmental Health