Ward's® Chemistry Forming Red, White and Blue Demonstration
Catalog # 470010-844
Supplier: Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Ward's® Chemistry Forming Red, White and Blue Demonstration
- Cat. no.:470010-844
- Includes:Instructions
- Supplier No.:WARD470010-844
- No. of Uses:7
Specifications
About this item
A Great Way to Introduce Chemical Reaction Types
- Provide for interesting discussion of 3 different chemical reactions
This “patriotic” demonstration illustrates complex ion formation, double replacement reaction and Prussian blue. Three reactions are carried out in adjacent beakers resulting in red, white and blue solutions.
Ordering information: Includes iron(III) chloride solution, potassium ferrocyanide solution, potassium thiocyanate solution, silver nitrate solution.