Wards® Coloring Solutions
Wards® Coloring Solutions
Supplier:   Ward's Science
Wards® Coloring Solutions
Supplier:  Ward's Science
Certificates
Wards® Coloring Solutions
Wards® Coloring Solutions
5 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Food colors in dropper bottles.
  • Concentrated color solutions
  • Blue, red, green, and yellow solutions available as a set or individually
  • Shelf life of three years
Product Family Options
Product Information
  • Taille
    Color
    Availability
    Price
  • 30 ml
    Blue
    Log in for availability
    Log in for availability
    Each
    Each - $7.05
    $7.05
    Each
       $7.05
    Supplier Number: FF0400-30ML
  • 30 ml
    Dark red
    Log in for availability
    Log in for availability
    Each
    Each - $6.80
    $6.80
    Each
       $6.80
    Supplier Number: AD-FF0441-30ML
  • 30 ml
    Green
    Log in for availability
    Log in for availability
    Each
    Each - $6.95
    $6.95
    Each
       $6.95
    Supplier Number: FF0420-30ML
  • 30 ml
    Yellow
    Log in for availability
    Log in for availability
    Each
    Each - $12.00
    $12.00
    Each
       $12.00
    Supplier Number: FF0460-30ML
  • Set of 4 Bottles, 50 mL Each
    Assorted
    Log in for availability
    Log in for availability
    Each
    Each - $54.65
    $54.65
    Each
       $54.65
    Supplier Number: FF0480-SET