Folding Digital Thermometer | Avantor
Folding Digital Thermometer
Folding Digital Thermometer, –50.00…+150 °C (–58.00…+302 °F)
Catalog # 470123-712
Fournisseur #:  PT300M
Folding Digital Thermometer
Catalog # 470123-712
Folding Digital Thermometer
Folding Digital Thermometer
Folding Digital Thermometer, –50.00…+150 °C (–58.00…+302 °F)
Price
$126.45
Log in pour la disponibilité
Each
Product Details & Documents
Large, Easy to Read Digital Display.
  • Measures in both C° and F°
  • Display turns on automatically when the probe is unfolded
  • Average response time 1 second
Stores the minimum and maximum temperatures.
Ordering information: Meter includes one AAA battery and protective sleeve.
Spécifications
  • Désignation:
    Folding Digital Thermometer
  • Range:
    –50.00…+150 °C (–58.00…+302 °F)