Flexible Probe
Flexible Probe, Stainless steel, 152 mm (6")
Catalog # 470007-696
Supplier: Dunrite Instruments
Supplier #: 140959
Jump to:
Product Details & Documents
Both ends of this flexible instrument are functional: the flat end with the eye can be threaded for ID tags, while other end with a thickened “nub” is useful for delicate probing without puncturing tissue.
Specifications
- Désignation:Flexible Probe
- Longueur:152 mm (6")
- Matière:Stainless steel