Flat Plastic Meter Stick
Flat metric meter stick, Plastic, 1 m
Catalog # 470047-182
Supplier: SI Manufacturing
Supplier #: 11050-A
Jump to:
Product Details & Documents
Flexible and flat, this durable plastic meter stick is marked in cm with 1 mm divisions on one side and with alternate-colored dm bars on the other, to illustrate the relationships between units.
Flexible and flat, this durable plastic meter stick is marked in cm with 1mm divisions on one side and with alternate-colored dm bars on the other, to illustrate the relationships between units.
Specifications
- Désignation:Flat metric meter stick
- Longueur:1 m
- Matière:Plastic