ContactUsLinkComponent
Flask Form Electroscope
  470020-542
undefined
Flask Form Electroscope
  470020-542
 :  4-50150
  • Description:
    Flask Form Electroscope
  • Cat. no.:
    470020-542
  • Supplier No.:
    7-501-5
Simple and Inexpensive
Use this compact apparatus contained in a 250 mL Pyrex flask. A nickel-plated rod with two aluminum leaves on one end fits into a rubber stopper in the neck of the flask. Charge the rod, and the leaves move apart.