To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Description:WARD’S Flame Test and Spectral Analysis Lab Activity
- Cat. no.:470012-260
- Supplier No.:WARD470012-260
An Effective Introduction to Spectral Analysis
Astronomers learn important information about the composition of stars by analyzing their spectra. To gain an understanding of this process, students use a spectroscope to observe and describe the spectra of four different chemical salts when heated by a flame. The kit comes with four salts, four nichrome wire test loops, six spectroscopes, a teacher’s guide, and student copymasters. It includes enough materials for 30 or more tests.