Fixed Right Angle Clamp Holders
Fournisseur: Shiv Dial Sud & Sons
Certificats
À propos de cet article
Perpendicular, V-Shaped Jaws.
- Thumbscrews 180° apart
- Hold support rods up to 3/4"" OD at a 90° angle.
This all brass clamp holder allows you to connect support rods together up to 19 mm in diameter.
Frequently Bought Together