About this item
Compare and contrast species from five kingdoms of life
- Arrows help students locate important features
- Mounted in a clear plastic holder that protects it on both sides
- Set of 15 includes detailed lesson plan on the topic under study
Explore the diversity of life all through the lens of the Microslide viewer as this set includes incredible images of organisms from all five Kingdoms of Life.
Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own Microslides. A detailed text folder is included. Viewers are available separately.
Contents:
Euglena, A classification puzzle (400X)
Kingdom Monera, Nitrogen-fixing bacteria (2,000X)
Kingdom Monera, Gloeocapsa(1,000X)
Kingdom Protista, Paramecium (100X)
Kingdom Protista, Spirogyra (150X)
Kingdom Fungi, Mushroom ( 1/2X and 400X)
Kingdom Plantae, Onion (110X) and Elodea (150X)
Kingdom Animalia, Liver cells (750X)
Box of 15 Sets also Includes:
Background subject information
Reproducible student worksheets with answer keys