Fish Mitosis Slide | Avantor
Fish Mitosis Slide
Fish Mitosis, Section, Weigert’s and Eosin
Catalog # 470182-554
Supplier:   Avantor
Fish Mitosis Slide
Catalog # 470182-554
Supplier:  Avantor
Fish Mitosis Slide
Fish Mitosis Slide
Fish Mitosis, Section, Weigert’s and Eosin
Price
$38.35
Log in pour la disponibilité
Each
Product Details & Documents
Explore mitosis and genetic structures
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
The finest slide for teaching mitotic division. Nine typical sections of a blastodisc from several different eggs to show many mitotic figures at various stages of mitosis.
Spécifications
  • Désignation:
    Fish Mitosis
  • Stain:
    Weigert’s and Eosin
  • Orientation:
    Section