Fish Mitosis Slide
Fish Mitosis, Section, Weigert’s and Eosin
Catalog # 470182-554
Supplier: Avantor
Jump to:
Product Details & Documents
Explore mitosis and genetic structures
- Identifying features clearly distinguishable
- Rigorous quality control standards
The finest slide for teaching mitotic division. Nine typical sections of a blastodisc from several different eggs to show many mitotic figures at various stages of mitosis.
Spécifications
- Désignation:Fish Mitosis
- Stain:Weigert’s and Eosin
- Orientation:Section