Ward's® Fingerprint Classification Chart | Avantor
Ward's® Fingerprint Classification Chart
WARD’S Fingerprint Classification Chart
Catalog # 470015-140
Supplier:   Avantor
Ward's® Fingerprint Classification Chart
Catalog # 470015-140
Supplier:  Avantor
Ward's® Fingerprint Classification Chart
Ward's® Fingerprint Classification Chart
WARD’S Fingerprint Classification Chart
Price
85,25 $
Log in pour la disponibilité
Each
Product Details & Documents
Learn How Forensic Scientists Classify Types
  • Size: 21 x 34"
Display detailed images and definitions of various fingerprint patterns on your walls for your students to study. The poster shows loops, arches, and whorls, as well as specific friction ridge characteristics such as bifurcation, enclosure, ridge endings, and more. Size: 21" x 34".
Spécifications
  • Désignation:
    WARD’S Fingerprint Classification Chart