Ward's® Finger Maze Set
WARD’S Finger Maze Set
Catalog # 470015-782
Supplier: Avantor
Jump to:
Product Details & Documents
Demonstrate the Process of Learning
This fun activity is performed on one of four different vacuum-formed maze boards and illustrates the concepts of goal insight, repetition, trial and error, and interference. It comes with an instruction manual. Size: 9 3/4" x 9 3/4".
Spécifications
- Désignation:WARD’S Finger Maze Set