Fine-Point Forceps With Guide Pins | Avantor
Fine-Point Forceps With Guide Pins
Supplier:   Dunrite Instruments
Fine-Point Forceps With Guide Pins
Certificates
Fine-Point Forceps With Guide Pins
Fine-Point Forceps With Guide Pins
2 Options Available Below
Product Details & Documents
Slip proof fine point forceps.
  • Guide pins keep tip aligned
  • Prevents slipping and twisting when closed
  • Textured tips improve hold on slippery material
Used in our own labs, these forceps feature a built-in guide pin to keep the fine tips aligned, which is important for grasping individual vessels or nerves. Stainless steel. Length: 4¹/₂".
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Longueur
    Tip Style
    Availability
    Price
  • Fine-Point Forceps With Guide Pin
    Stainless steel
    115 mm (4.5")
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 6,25 $
    6,25 $
    Log in pour la disponibilité
    Box of 50
    Box of 50 - 293,55 $
    293,55 $
    Each
       6,25 $
    Box of 50
       293,55 $
    Supplier Number: 140541
  • Fine-Point Forceps With Guide Pin
    Stainless steel
    115 mm (4.5")
    Curved
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 7,05 $
    7,05 $
    Log in pour la disponibilité
    Box of 50
    Box of 50 - 334,95 $
    334,95 $
    Each
       7,05 $
    Box of 50
       334,95 $
    Supplier Number: 140531