Ficus, Leaf Slide
Ficus, Leaf, Cross section, Quadruple
470181-322
: Avantor
Demonstrates Leaf Characteristics
- Identifying features clearly distinguishable
- Rigorous quality control standards
Rubber plant. Showing prominent palisade layer, multiple epidermis, and sunken stomata.
- Désignation:Ficus, Leaf
- Stain:Quadruple
- Orientation:Cross section