Fast Release Pi-Pumps | Avantor
Fast Release Pi-Pumps
Supplier:   Bel-Art
Fast Release Pi-Pumps
Supplier:  Bel-Art
Certificates
Fast Release Pi-Pumps
Fast Release Pi-Pumps
1 Options Available Below
Product Details & Documents
Designed for fast and accurate one-handed operation, this pump will reduce the risk of injury from repetitive pipetting.
  • Each pump is resistant to acids, bases, and most solvents
  • Disassembly for cleaning is easy
The fast-release trigger expels the entire contents at one shot, or you can use the thumb wheel for precision dispensing.
Product Family Options
Informations sur le produit
  • Désignation
    Capacité
    Color
    Availability
    Price
  • Fast Release Pi-Pump
    2 ml
    Blue
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $103.00
    $103.00
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $1,236.00
    $1,236.00
    Each
       $103.00
    Case of 12
       $1,236.00
    Supplier Number: F37904-0002
  • Fast Release Pi-Pump
    10 ml
    Discontinued
    Supplier Number: F37904-0010
  • Fast Release Pi-Pump
    25 ml
    Discontinued
    Supplier Number: F37904-0025