Eyepiece Pointer Wire
Eyepiece Pointer Wire
470021-924
: PT101
The pointer is housed in a disc that fits into your microscope's eyepiece, and is held in place by a spring. Point out specific structures under the microscope.
- Désignation:Eyepiece Pointer Wire