Exploring With a Microscope II Slide Set
Exploring With a Microscope II Slide Set, 12
Catalog # 470183-640
Supplier: Avantor
Jump to:
Product Details & Documents
Includes an Illustrated Study Guide Highlighting Microscopic Features of “Common Things”
- All identifying features are clearly distinguishable
- Rigorous quality control standards
- In-house experts available to answer your questions
View familiar objects and materials at the microscopic level. All slides are whole mounts. This set comes with an illustrated study guide.
Ordering information: Set includes Quartz sand; Cosmetic types; Dandruff; Cornstarch; Salt crystals; Nylon and Wool Fibers; Toothpaste; Human Hair; Sawdust; Iron rust, Common dust and Chalk.
Spécifications
- Désignation:Exploring With a Microscope II Slide Set
- No. of Slides:12