Ward's® Exploring Mechanical Weathering Lab Activity | Avantor
Ward's® Exploring Mechanical Weathering Lab Activity
Supplier:   Avantor
Ward's® Exploring Mechanical Weathering Lab Activity
Supplier:  Avantor
Certificates
Ward's® Exploring Mechanical Weathering Lab Activity
Ward's® Exploring Mechanical Weathering Lab Activity
2 Options Available Below
Product Details & Documents
The Exploring Mechanical Weathering Lab Activity Shows the weathering of rocks on the Earth’s surface.
  • Students observe how the environment effects rocks
  • Developed by Ward's geologists
  • Teacher's guide included
  • Aligned to NGSS standards (MS/HS: ESS1, ESS2)
It’s easy to recreate the process of mechanical weathering in the lab. Students can observe what happens to different rock samples when each is shaken and abraded in a container of water.
Their findings help them understand the various factors that influence mechanical weathering. The lab includes a teacher’s guide and student copymasters.
The kit is also available for purchase with a link to a NewPath digital lesson, which includes a variety of resources on the topic, including background information, illustrations and self-assessments.
Product Family Options
Informations sur le produit
  • Désignation
    No. of Lab Groups
    Availability
    Price
  • Ward's® Exploring Mechanical Weathering Lab Activity
    1
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Kit of 1
    Kit of 1 - $109.80
    $109.80
    Log in pour la disponibilité
    Pallet of 100
    Pallet of 100 - $10,430.35
    $10,430.35
    Kit of 1
       $109.80
    Pallet of 100
       $10,430.35
  • Ward's® Exploring Mechanical Weathering Lab Activity
    10
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    KIT(1items)
    KIT(1items) - $500.19
    $500.19
    Log in pour la disponibilité
    Kit of 1
    Kit of 1 - $681.10
    $681.10
    Log in pour la disponibilité
    Pallet of 15
    Pallet of 15 - $9,705.90
    $9,705.90
    KIT(1items)
       $500.19
    Kit of 1
       $681.10
    Pallet of 15
       $9,705.90
  • Kit with NewPath Digital Resources
    1
    Discontinued