Exploring Magnets Kit
Exploring Magnets Kit
Catalog # 470012-626
Supplier: Science First
Fournisseur #: 615-8510
Jump to:
Product Details & Documents
Unique Shapes Challenge Students’ Assumptions
- Sealed viewer for clean learning
Actually view magnetic field lines with this unique magnetic viewer and set of seven very powerful neodymium rare earth magnets. A bar, star, heart, disk, triangle, ring, and ball magnet are included. The sealed viewer safely contains iron fillings that demonstrate the different magnetic field orientations of both common and odd shaped magnets.
Spécifications
- Désignation:Exploring Magnets Kit