ContactUsLinkComponent
Ward's® Exploring Environmental Science: Water Quality Testing and Analysis CD-ROM
Ward's® Exploring Environmental Science: Water Quality Testing and Analysis CD-ROM
Catalog # 470029-004
Supplier:  JAROSLAW KUDAY
undefined
Ward's® Exploring Environmental Science: Water Quality Testing and Analysis CD-ROM
Catalog # 470029-004
Supplier:  JAROSLAW KUDAY
Supplier Number:  745097
Specifications
  • Description:
    WARD’S Exploring Environmental Science: Water Quality Testing and Analysis CD-ROM, Mac/Windows CD-ROM
  • Cat. no.:
    470029-004
  • Supplier No.:
    745097
Specifications
About this item
Multimedia Presentation on Water Sampling, Testing, and Analysis
This ”how-to“ guide on CD-ROM uses photographs, illustrations, video clips, and text with hyperlinks to more information to demonstrate several water sampling and testing techniques. The program demonstrates how to correctly use instruments that measure dissolved oxygen, turbidity, and conductivity, as well as other testing apparatus. It also features extensive background information on aquatic communities, microlife bioindicators, and water pollutant limits and their effects on the environment. A separate section includes suggestions for hands-on lab investigations, safety guidelines, and a bibliography.
System Requirements
Mac: 68040 processor or higher, System 7.0 or higher, 16MB RAM
Windows: 486/50 processor or higher, Windows 3.1 or Windows 95, 16MB RAM, sound card recommended