ContactUsLinkComponent
Ward's® Chemistry Exothermic and Endothermic Reaction Demonstrations
Ward's® Chemistry Exothermic and Endothermic Reaction Demonstrations
Catalog # 470013-162
Supplier:  Avantor
undefined
Ward's® Chemistry Exothermic and Endothermic Reaction Demonstrations
Catalog # 470013-162
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Ward's® Chemistry Exothermic Reaction Demonstration
  • No. of Lab Groups:
    1
  • Cat. no.:
    470013-162
  • Includes:
    Instruction Manual
  • Supplier No.:
    WARD470013-162
Specifications
About this item
Tactile Demonstrations of a Chemical Reaction.
  • Takes approximately 5 minutes to complete each demonstration
  • Includes all required materials
In these demonstrations, exothermic and endothermic reactions are safely carried out inside a plastic bag so students can pass the bag around and feel the results of the reaction. For the exothermic reaction the bag will become warm to the touch, while in the endothermic reaction the bag will become cold to the touch. Each kit contains all the materials necessary for five demonstrations prepackaged in the plastic bags.