Euglena Slide
Euglena, Whole Mount, Fast Green and Mallory's
Catalog # 470181-458
Supplier:   Avantor
Euglena Slide
Catalog # 470181-458
Supplier:  Avantor
Euglena Slide
Euglena Slide
Euglena, Whole Mount, Fast Green and Mallory's
Price
$9.50
Log in pour la disponibilité
Each
Product Details & Documents
Single Celled Flagellates
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Large euglenoid which shows discoid chromatophores, paramylon bodies, and stigma.
Spécifications
  • Désignation:
    Euglena
  • Stain:
    Fast Green and Mallory's
  • Orientation:
    Whole Mount
Accessories
Informations sur le produit
  • Catalog #
    Media
    No. of Students
    Availability
    Price
  • Euglena sp. Study Set
    Euglena sp. Study Set
    470189-298
    Euglena medium
    30
    Log in pour la disponibilité
    Kit of 1 - 30,00 $
Informations sur le produit
  • Catalog #
    Media
    No. of Students
    Availability
    Price
  • Euglena sp (Large)
    Euglena sp (Large)
    470180-112
    Euglena medium
    100
    Log in pour la disponibilité
    Each - 47,30 $
Informations sur le produit
  • Catalog #
    Media
    No. of Students
    Availability
    Price
  • Black Planaria Culture (Small)
    Black Planaria Culture (Small)
    470176-452
    Dechlorinated water
    30
    Log in pour la disponibilité
    Each - 28,35 $
Informations sur le produit
  • Catalog #
    Media
    No. of Students
    Availability
    Price
  • Euglena gracilis (Small)
    Euglena gracilis (Small)
    470175-818
    Euglena medium
    30
    Log in pour la disponibilité
    Each - 15,70 $