Euglena
Catalog # 470177-496
Supplier: Avantor
Specifications
- Description:Euglena
- Cat. no.:470177-496
- Stain:Schneiders and Fast Green
- Supplier No.:WARD470177-496
- Orientation:Whole Mount
Specifications
About this item
Explore this often studied flagellate
- Identifying features clearly distinguishable
- Rigorous quality control standards
General study. Demonstrates nucleus, chloroplasts, contractile vacuole, and flagellum.