Etherizer
Etherizer, Drosophila, w/ Dropper Pipet
470154-018
: Avantor
Specially designed unit to safely and easily etherize Drosophila or other living materials.
- Etherized Drosophila drop into a collecting bottle
The bottle containing live material is inverted over the funnel. A few drops of ether are placed on a dispersing wick under the rubber plug.
: Includes dropper pipet.
- Désignation:Etherizer, Drosophila, w/ Dropper Pipet