ContactUsLinkComponent
Essential Physics Demo: Large Refraction Block
Essential Physics Demo: Large Refraction Block
Catalog # 470018-086
Small Business Enterprise
undefined
Essential Physics Demo: Large Refraction Block
Catalog # 470018-086
Supplier Number:  1808-61
Small Business Enterprise
Specifications
  • Description:
    Essential Physics Demo: Large Refraction Block
  • Cat. no.:
    470018-086
  • Supplier No.:
    1808-61
Specifications
About this item
The exciting demonstrations connect students to core physics and physical science concepts.
  • Large-Scale Demonstration
  • Simple Set Up and Operation
  • Instructions Included
The exciting demonstrations connect students to core physics and physical science concepts. With quick assembly and operation, these demos give educators more time to work with students.
Visually display the concepts of reflection and refraction by simply shining a laser pointer towards this acrylic block. The kit includes the block and demonstration instructions.