Order Entry
Canada
ContactUsLinkComponent
Erada-Stain Biological/Chemical Stain Remover
Catalogue # 470121-812
undefined
Erada-Stain Biological/Chemical Stain Remover
Catalogue # 470121-812
Supplier Number:  206
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Spécifications

  • Description:
    Erada-Stain Biological/Chemical Stain Remover, Tube
  • Size:
    170 g (6 oz.)
  • Cat. no.:
    470121-812
  • Supplier No.:
    206

Spécifications

À propos de cet article

Erada-Stain® is a very mild cream cleaner / stain remover that can be used on hands or lab equipment.

Rapidly removes Wright’s stain and most medical and histological stains from hands, glassware and laboratory equipment. Stains disappear in seconds when applied. Rinse with water. Cream is available in 170 g squeeze tube.