Equilateral Prisms
Equilateral Prisms
Equilateral Prisms
Certificates
Equilateral Prisms
Equilateral Prisms
6 Options Available Below
Product Details & Documents
For all Optical Applications
  • These prisms are polished on all faces
  • They are made of shatter-resistant acrylic material or optical quality glass
These prisms are polished on all faces. They are made of shatter-resistant acrylic material or optical quality glass. The length of the sides and the height are given for each prism.
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • Equilateral Prisms, Glass, 25 x 25 mm
    Log in for availability
    Log in for availability
    Each
    Each - $36.65
    $36.65
    Each
       $36.65
    Supplier Number: PGE025
  • Equilateral Prisms, Acrylic, 25 x 12 mm
    Log in for availability
    Log in for availability
    Each
    Each - $1.55
    $1.55
    Clearance
    Each
       $1.55
    Supplier Number: PH0554A
  • Equilateral Prisms, Acrylic, 50 x 20 mm
    Log in for availability
    Log in for availability
    Each
    Each - $18.95
    $18.95
    Each
       $18.95
    Supplier Number: PH0554B
  • Equilateral Prisms, Glass, 25 x 50 mm
    Log in for availability
    Log in for availability
    Each
    Each - $3.00
    $3.00
    Clearance
    Each
       $3.00
    Supplier Number: PH0554D
  • Equilateral Prisms, Glass, 25 x 75 mm
    Log in for availability
    Log in for availability
    Each
    Each - $5.45
    $5.45
    Clearance
    Each
       $5.45
    Supplier Number: PH0554E
  • Equilateral Prisms, Glass, 25 x 100 mm
    Log in for availability
    Log in for availability
    Each
    Each - $54.35
    $54.35
    Each
       $54.35
    Supplier Number: PGE100