Enzyme Analysis Activity Kit
Enzyme Analysis Activity Kit
Catalog # 470149-874
Supplier: G-Biosciences
Supplier #: BE-405
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Study Four Different Enzyme Concepts
This flexible, hands-on kit teaches enzyme kinetic and regulation of enzyme activity, role of substrate concentration, effects of pH and temperature on reaction rate, and inhibition of enzyme reaction. Materials provided for six groups of up to five students each.
Specifications
- Désignation:Enzyme Analysis Activity Kit