ContactUsLinkComponent
Emergency Drench Shower
Catalog # 470016-870
undefined
Emergency Drench Shower
Catalog # 470016-870
Supplier Number:  01-0530-01
Specifications
  • Description:
    Emergency Drench Shower
  • Cat. no.:
    470016-870
  • Supplier No.:
    01-0530-01
Specifications
About this item
Safety Feature No Laboratory Should Be Without.
  • Proper plumbing installation and connections required
  • Includes all necessary parts and instructions
Whether or not it is mandated by your school district, emergency drench showers are a safety feature no laboratory should be without. This shower meets ANSI regulations, with user friendly features including easily accessed pull valves that stay open until manually closed and highly visible yellow plastic shower heads designed to deliver a concentrated flood of water in a drench column at least 20" in Ø. These are unregulated shower heads with 30 to 80 psi dynamic water pressure. At 30 to 40 it runs at 53 gallons per minute, at 50 to 60 it runs at 54 gallons per minute, and at 70 to 80 it runs at 55 gallons per minute. Constructed of heavy galvanized steel 1 ¹/₄" piping.