Ward's® ELISA Simulation Lab Activity | Avantor
Ward's® ELISA Simulation Lab Activity
Supplier:   Avantor
Ward's® ELISA Simulation Lab Activity
Supplier:  Avantor
Certificates
Ward's® ELISA Simulation Lab Activity
Ward's® ELISA Simulation Lab Activity
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Safe and Colorful Activity
  • No refrigeration or expensive antibodies required
  • Teacher developed and classroom tested
  • Simulates the test for bacterial infection
  • Time Required: 45 minutes
  • Aligned to NGSS standards (HS: PS1, LS1)
Biology teacher, Brenda Royal, developed a quick and easy way to introduce concepts in immunology. Students perform a simulated ELISA test on fluid samples from two hypothetical patients to determine if either has been infected with an emerging bacterial strain. Materials for 10 student groups are included, along with teacher and student instructions.
Product Family Options
Informations sur le produit
  • Désignation
    No. of Lab Groups
    Availability
    Price
  • ELISA simulation kit
    1
    Log in pour la disponibilité
    Log in pour la disponibilité
    Kit of 1
    Kit of 1 - $94.65
    $94.65
    Kit of 1
       $94.65
  • ELISA simulation kit
    10
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $197.95
    $197.95
    Log in pour la disponibilité
    Pallet of 36
    Pallet of 36 - $6,770.15
    $6,770.15
    Each
       $197.95
    Pallet of 36
       $6,770.15