Vernier® Electrostatics Kit
Vernier electrostatics kit
Catalog # CA470017-514
New Product
Supplier: VERNIER CANADA SCIENCE EDUCATION IN
Supplier #: ESK-CRG
Vernier® Electrostatics Kit
Catalog # CA470017-514
Supplier: VERNIER CANADA SCIENCE EDUCATION IN
New Product
Jump to:
Product Details & Documents
Inclusive package for inclusive study.
- Using Faraday’s Ice Pail
- Production of charge by friction
- Transfer of charge by contact
- Charging by Induction
- Quantitative and qualitative measurement of charge
An accompaniment to the Vernier Charge sensor, this kit enables the inquiry and examination into a wide range of experiments in electrostatics. Measure charge in a quantitative and qualitative manner, and see how friction, induction, and the contact of two objects can vastly affect its charge. The kit includes everything you need for deep exploration- a Faraday pail and cage, grounding wires and plane, a wrist strap, a proof plane, and the following charge producers: wool, vinyl, a PVC rod, a nylon rod, and cotton cloth.
For educational use only
Specifications
- Désignation:Vernier electrostatics kit