ContactUsLinkComponent
Electrostatic Kit
Catalog # 470120-700
Supplier:  Wards
undefined
Electrostatic Kit
Catalog # 470120-700
Supplier:  Wards
Supplier Number:  PH0900
Specifications
  • Description:
    Electrostatic Kit
  • Cat. no.:
    470120-700
  • Supplier No.:
    PH0900
Specifications
About this item
Detect the Presence of Static Electricity
  • User Friendly
  • Reliable Materials
Charge electroscopes, show electrostatic attraction, and more with the contents of this activity kit. A simple electroscope is included for easily identifying static electricity. A series of other materials including metalized polystyrene spheres, polyethylene tiles, aluminum cans, nylon, PVC, wire, rubber cloth, and wire hook are also supplied for completing electricity experiments.