Electronics Snap Circuits Kits | Avantor
Electronics Snap Circuits Kits
Supplier:   Elenco Electronics
Electronics Snap Circuits Kits
Supplier:  Elenco Electronics
Certificates
Electronics Snap Circuits Kits
Electronics Snap Circuits Kits
1 Options Available Below
Product Details & Documents
Electronics Snap Circuit Kits feature easy-to-connect components, making them perfect for elementary electronics labs.
  • Easy-To-Connect Components
  • Instructions Included
  • Color-Coded Parts
  • Designed for Students New to Circuit Assembly
  • Projects Include Switch Circuits through Motors, Logic Circuits, and More
The circuit components are firmly attached to plastic bases, color coded by function, and mounted on snap bases for easy assembly and reliable contact.
When laid out on the base, the functioning circuit resembles the conventional circuit diagram, so students can relate the actual circuit to the description of its function. Projects range from simple switch circuits through motors to logic circuits and IC-controlled devices.
Ordering information: The kits contain a plastic base plate with molded bases for arranging the circuits, the necessary electronic components and connectors, an illustrated instruction manual, and a compartmented storage box for easy checking. Two AA-size batteries (not included) provide power.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Electronics Snap Circuits Kits, Jr. 100 Experiments Kit
    Log in pour la disponibilité
    Log in pour la disponibilité
    Kit of 1
    Kit of 1 - $78.05
    $78.05
    Kit of 1
       $78.05
    Supplier Number: SC100
  • Snap Circuits Jr. Kit
    Discontinued
    Supplier Number: SC-100
  • Discontinued
    Supplier Number: -