Electric Gyrospinner
Catalog # 470110-846
Supplier: COOK IRON STORE
Specifications
- Description:Electric Gyrospinner
- Cat. no.:470110-846
- Supplier No.:100-N/7
Specifications
About this item
Spins Your Gyroscope to High Speeds Quickly and Reliably.
- This gyrospinner turns at 5,000 and 10,000 rpm and can be used to spin all your demonstration gyroscopes
The electric gyrospinner consists of a special universal motor with a mandrel and driving disk and a molded rubber O-ring mounted on the shaft. Note: Must remove abrasive cover from the spindle before use.