To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Description:Effects of Drugs on Daphnia Kit
- No. of Lab Groups:15
- Cat. no.:470018-280
Simple, Effective Investigation Uses Living Specimens
- Pre and post-laboratory assessments
- Links to real world concepts
- Estimated class/lab time required: 40-60 minutes
Students use WARD’S new method of magnifying cells and Daphnia netting to view the Daphnia’s heartbeat. Students will familiarize themselves with some common stimulants and depressants by testing four different compounds and analyzing the results of these substances on the Daphnia’s heartrate. By observing how drugs affect a living organism, it becomes easier for students to correlate the effects of drugs on their own bodies. You get enough materials for 15 setups, a teacher’s guide, and student copy master. Note: Coupon included for perishable materials. Redeem by mail, fax, phone, or e-mail.