Milwaukee® Digital Salinity Refractometer | Avantor
Milwaukee® Digital Salinity Refractometer
Digital salinity refractometer, 0 - 50 PSU: 0 - 150 ppt, 1.000 - 1.114 S.G. (20/20), 0...80 °C / 32...176 °F, ±2 PSU, ±2 ppt , ±0.002 S.G. (20/20), ±0.3 °C (±0.5 °F)
Catalog # 470017-304
Fournisseur #:  MA887
Milwaukee® Digital Salinity Refractometer
Catalog # 470017-304
Milwaukee® Digital Salinity Refractometer
Milwaukee® Digital Salinity Refractometer
Digital salinity refractometer, 0 - 50 PSU: 0 - 150 ppt, 1.000 - 1.114 S.G. (20/20), 0...80 °C / 32...176 °F, ±2 PSU, ±2 ppt , ±0.002 S.G. (20/20), ±0.3 °C (±0.5 °F)
Price
$384.00
Log in pour la disponibilité
Each
Product Details & Documents
Ideal for student laboratories with minimal mess – only 2 to 3 drops needed and results appear instantly on a digital display.
  • This durable refractometer is easy to use and requires minimal maintenance Fast, accurate results with digital dual level LCD readout Automatically temperature compensated Simple one-point calibration using distilled or deionized water
Want accurate, reliable and repeatable results? This Milwaukee Refractometer makes learning hands on simple with fast, accurate readings with just a few drops – perfect for food science, chemistry, and beverage experiments in the classroom or laboratory.
Ordering information: Batteries included. Does NOT include distilled or deionized water for calibration.
Spécifications
  • Désignation:
    Digital salinity refractometer
  • Accuracy:
    ±2 PSU, ±2 ppt , ±0.002 S.G. (20/20), ±0.3 °C (±0.5 °F)
  • Range:
    0 - 50 PSU: 0 - 150 ppt, 1.000 - 1.114 S.G. (20/20), 0...80 °C / 32...176 °F