Echinococcus sp., Hydatid Slide
Echinococcus sp., Hydatid, Whole mount, Schneider's and Fast Green
Catalog # 470182-092
Supplier: Avantor
Jump to:
Product Details & Documents
Intestinal Parasite
- Identifying features clearly distinguishable
- Rigorous quality control standards
Shows hydatid sand, fluid of hydatid cyst, liberated blood capsules, and free scolices.
Specifications
- Désignation:Echinococcus sp., Hydatid
- Stain:Schneider's and Fast Green
- Orientation:Whole mount