<i>Echinococcus sp</i>., Hydatid Slide
Echinococcus sp., Hydatid Slide
Echinococcus sp., Hydatid, Whole mount, Schneider's and Fast Green
Catalog # 470182-092
Supplier:   Avantor
<i>Echinococcus sp</i>., Hydatid Slide
Catalog # 470182-092
Supplier:  Avantor
<i>Echinococcus sp</i>., Hydatid Slide
Echinococcus sp., Hydatid Slide
Echinococcus sp., Hydatid, Whole mount, Schneider's and Fast Green
Price
$18.15
Log in for availability
Each
Product Details & Documents
Intestinal Parasite
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Shows hydatid sand, fluid of hydatid cyst, liberated blood capsules, and free scolices.
Specifications
  • Désignation:
    Echinococcus sp., Hydatid
  • Stain:
    Schneider's and Fast Green
  • Orientation:
    Whole mount