Vernier® EasyLink interface
Vernier easyLink interface
CA470017-138
New Product
: EZ-LINK
Inexpensive data collection direct to any TI-Nspire® handheld or TI-84 Plus graphing calculator.
- Connects BTA (sensor) to mini-A USB (calculator)
- Compatible with all models of TI-84 Plus, TI-84 Plus CE, and TI-Nspire® CXII Texas Instruments calculators
- Supports over 62 analog Vernier sensors (Digital sensors and microphone are incompatible)
EasyLink® is a single-channel interface that plugs into the USB port of a TI-84 Plus graphing Calculator or TI-Nspire® Handheld. Use EasyLink® and a supported sensor to explore math and science concepts found in real-world phenomena such as light, pressure, force, and more.
- Désignation:Vernier easyLink interface