Easy Cutter
Easy Cutter
Catalog # 470100-816
Supplier: MIDWEST PRODUCTS
Fournisseur #: 1126
Jump to:
Product Details & Documents
Perfect for cutting balsa for bridge building and other projects.
- Textured grip for comfort
- Double ground, heat treated high carbon steel blade stays sharp longer
- Cutting plate marked in English and metric units
- Clearly defined 45°, 60°, 90° and 120° angles
Cuts up to 1/2" thick strips of wood but is best used for applications requiring materials up to 1/4" thick. Other features include a built-in bottle opener and safety latch.
Spécifications
- Désignation:Easy Cutter