Earthworm Dissection Lab | Avantor
Earthworm Dissection Lab
Lab Activity, Boreal, Earthworm Dissection, Junior High
Catalog # 470044-554
Supplier:   Avantor
Earthworm Dissection Lab
Catalog # 470044-554
Supplier:  Avantor
Earthworm Dissection Lab
Earthworm Dissection Lab
Lab Activity, Boreal, Earthworm Dissection, Junior High
Price
$94.24
Vente
Valable jusqu'au 31/12/9999
Log in pour la disponibilité
Kit of 1 Pallet of 36
Product Details & Documents
With this Complete Earthworm Dissection Lab, you’ll receive: 10 high quality specimens that are easy to pin and dissect, packed in formaldehyde-free Borealene II. 20 student manuals with pre-test, post-test and easy to follow step-by-step instructions. 10 disposable dissection trays that offer direct pinning to the surface with no cleanup or resurfacing. Teacher’s guide with student objectives, teaching hints and answers to student tests. Replacement Student Manuals and Teacher’s Guides may also be purchased separately.
Spécifications
  • Désignation:
    Lab Activity, Boreal, Earthworm Dissection, Junior High
Accessories
Informations sur le produit
  • Catalog #
    Grade
    Size
    Availability
    Price
  • WARDSafe™
    WARDSafe™
    470233-546
    Laboratory
    19 L
    Log in pour la disponibilité
    Each (19l) - $345.85