Duodenum Slide
Duodenum, Cross section, Hematoxylin and Eosin
Catalog # 470177-614
Supplier: Avantor
Jump to:
Product Details & Documents
Explore a section of the digestive system
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Shows Brunner’s glands and intestinal glands, known as Lieberkuhn’s crypts.
Spécifications
- Désignation:Duodenum
- Stain:Hematoxylin and Eosin
- Orientation:Cross section