Duct Tape | Avantor
Duct Tape
Duct Tape
Catalog # 470149-902
Supplier:   xpedx
Fournisseur #:  10070278
Duct Tape
Catalog # 470149-902
Supplier:  xpedx
Duct Tape
Duct Tape
Duct Tape
Price
29,45 $
Log in pour la disponibilité
Roll of 1
Product Details & Documents
Extra strength, non-elastic silver duct tape. Water, vapor, and flame resistant.
Packaging: Supplied as 54 m roll.
Spécifications
  • Désignation:
    Duct Tape