Ward's® Dual Magnifier
Fournisseur: Skywill Far East Industries
Certificats
À propos de cet article
This dual magnifier has two lenses for varying magnification.
- Two-in-one durable plastic magnifier
- 3X and 6X
- Aligned with GLOBE protocols
View specimens two different ways, with a two-in-one durable plastic magnifier: under 3X with 1 5/8" diameter lens, or 6X with 5/8" diameter lens.
Dimensions: 4 1/2L x 2"W
Frequently Bought Together