ContactUsLinkComponent
Drosophila Mutations Slide Set
Drosophila Mutations Slide Set
Catalog # 470183-514
Supplier:  Avantor
undefined
Drosophila Mutations Slide Set
Catalog # 470183-514
Supplier:  Avantor
Specifications
  • Type:
    Cellular Development
  • Description:
    Drosophila Mutations Slide Set
  • No. of Slides:
    9
  • Cat. no.:
    470183-514
  • Supplier No.:
    WARD470183-514
Specifications
About this item
Investigate Drosophila mutations and learn about genetic variability
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Prepared in Ward's® own labs by skilled technicians, the slides show the detailed anatomy of organisms commonly used in genetic and developmental studies.All slides are whole mounts.
Ordering information: Set includes Life cycle composite; Eye: Bar; White; Wing: Vestigial; Dumpy; Apterous; Body: Ebony; Homeotic: Aristapedia; and Antennapedia.