Down Feather Slide
Down Feather, Whole mount, Unstained
Catalog # 470181-826
Supplier: Avantor
Jump to:
Product Details & Documents
Illustrates Down Feather Structure
- Identifying features clearly distinguishable
- Rigorous quality control standards
Fine feathers found underneath other body feathers. They serve as padding and insulation.
Spécifications
- Désignation:Down Feather
- Stain:Unstained
- Orientation:Whole mount